ENA413164652 Enamine

5H,6H,7H-pyrrolo[1,2-a]imidazole-3-sulfonamide

  • MDL number MFCD27920002
  • Cas Number : 1797604-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-117719

Safety Information

msds https://www.enaminestore.com/catalog/EN300-117719
Availability details for
Quantity
  • Availability
  • Add to Cart