ENA413164659 Enamine

4-(2-bromoethyl)-1lambda6-thiomorpholine-1,1-dione hydrobromide

  • MDL number MFCD27920010
  • Cas Number : 1803583-00-7
  • Molecular Weight : 323.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-bromoethyl)thiomorpholine 1,1-dioxide hydrobromide
Inchi Code 1S/C6H12BrNO2S.BrH/c7-1-2-8-3-5-11(9,10)6-4-8;/h1-6H2;1H
Inchi Key AIYPYKOCNZNZKU-UHFFFAOYSA-N

Description

Purity 95
Salt Data Br
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-124668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-124668
Availability details for
Quantity
  • Availability
  • Add to Cart