ENA413164669 Enamine

[2-(2-fluoro-6-nitrophenyl)ethenyl]dimethylamine

  • MDL number MFCD26935947
  • Cas Number : 1807921-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-126980

Safety Information

msds https://www.enaminestore.com/catalog/EN300-126980
Availability details for
Quantity
  • Availability
  • Add to Cart