ENA413164671 Enamine

3-(dimethylsulfamoyl)propyl benzoate

  • MDL number MFCD26935949
  • Cas Number : 1803593-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-128848

Safety Information

msds https://www.enaminestore.com/catalog/EN300-128848
Availability details for
Quantity
  • Availability
  • Add to Cart