ENA413164672 Enamine

3-(furan-2-yl)-3-{[(4-methoxyphenyl)methyl]amino}propan-1-ol

  • MDL number MFCD27920019
  • Cas Number : 1803607-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129137
Availability details for
Quantity
  • Availability
  • Add to Cart