ENA413164676 Enamine

1-(oxan-4-yl)-3-(piperazin-1-yl)piperidin-2-one

  • MDL number MFCD26935950
  • Cas Number : 1803561-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129773
Availability details for
Quantity
  • Availability
  • Add to Cart