ENA413164683 Enamine

2-methoxy-N-[3-(piperidin-3-yl)-1H-pyrazol-4-yl]acetamide

  • MDL number MFCD26935953
  • Cas Number : 1803570-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-129903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129903
Availability details for
Quantity
  • Availability
  • Add to Cart