ENA413164694 Enamine

N-benzyl-3-(trifluoromethyl)oxolan-3-amine

  • MDL number MFCD26935957
  • Cas Number : 1638269-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-131723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-131723
Availability details for
Quantity
  • Availability
  • Add to Cart