ENA413164697 Enamine

3-ethyl-[1,2]oxazolo[5,4-b]pyridine-5-sulfonamide

  • MDL number MFCD26935959
  • Cas Number : 1803605-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-132452

Safety Information

msds https://www.enaminestore.com/catalog/EN300-132452
Availability details for
Quantity
  • Availability
  • Add to Cart