ENA413164701 Enamine

ethyl 2-(7-{[(tert-butoxy)carbonyl]amino}-4,5,6,7-tetrahydro-2H-indazol-2-yl)acetate

  • MDL number MFCD27920029
  • Cas Number : 1797646-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133375

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133375
Availability details for
Quantity
  • Availability
  • Add to Cart