ENA413164702 Enamine

tert-butyl 4-[1-(oxan-4-yl)-2-oxopiperidin-3-yl]piperazine-1-carboxylate

  • MDL number MFCD26935961
  • Cas Number : 1803560-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133406

Safety Information

msds https://www.enaminestore.com/catalog/EN300-133406
Availability details for
Quantity
  • Availability
  • Add to Cart