ENA413164707 Enamine

methyl 4-[3-(trifluoromethyl)phenyl]butanoate

  • MDL number MFCD26935966
  • Cas Number : 191155-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-133945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-133945
Availability details for
Quantity
  • Availability
  • Add to Cart