ENA413164715 Enamine

pyrimidine-4,5-diamine hydrochloride

  • MDL number MFCD27500670
  • Cas Number : 97846-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135258
Availability details for
Quantity
  • Availability
  • Add to Cart