ENA413164716 Enamine

[3-(trifluoromethyl)-1H-1,2,4-triazol-5-yl]methanamine hydrochloride

  • MDL number MFCD27500671
  • Cas Number : 1609395-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167...169
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135491

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-135491
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart