ENA413164721 Enamine

2-amino-2-(1-methylcyclohexyl)acetic acid hydrochloride

  • MDL number MFCD27500672
  • Cas Number : 1796905-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135717

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-135717
Availability details for
Quantity
  • Availability
  • Add to Cart