ENA413164722 Enamine

tert-butyl 3-[(2-methyl-1,3-thiazol-4-yl)methoxy]pyrrolidine-1-carboxylate

  • MDL number MFCD26935977
  • Cas Number : 1803571-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135832
Availability details for
Quantity
  • Availability
  • Add to Cart