ENA413164725 Enamine

1-benzyl-1H-1,2,3-triazol-4-amine hydrochloride

  • MDL number MFCD27920035
  • Cas Number : 1803586-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123...125
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136010

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136010
Availability details for
Quantity
  • Availability
  • Add to Cart