ENA413164727 Enamine

2,3-dihydro-1-benzoxepine-4-sulfonyl chloride

  • MDL number MFCD26935978
  • Cas Number : 1602704-82-4
  • Molecular Weight : 244.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dihydrobenzo[b]oxepine-4-sulfonyl chloride
Inchi Code 1S/C10H9ClO3S/c11-15(12,13)9-5-6-14-10-4-2-1-3-8(10)7-9/h1-4,7H,5-6H2
Inchi Key SIQCEPFBSLNZHE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-136173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136173
Availability details for
Quantity
  • Availability
  • Add to Cart