ENA413164728 Enamine

1-(3-methoxypropanesulfonyl)piperidin-4-yl methanesulfonate

  • MDL number MFCD26935979
  • Cas Number : 1803591-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136277
Availability details for
Quantity
  • Availability
  • Add to Cart