ENA413164732 Enamine

methyl 4-(2-cyanoethyl)morpholine-3-carboxylate

  • MDL number MFCD27500675
  • Cas Number : 1564865-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136680

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-136680
Availability details for
Quantity
  • Availability
  • Add to Cart