ENA413164734 Enamine

ethyl 2-(1-methyl-1H-pyrazole-4-carbonyloxy)-3-oxobutanoate

  • MDL number MFCD26935984
  • Cas Number : 1803586-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-136996
Availability details for
Quantity
  • Availability
  • Add to Cart