ENA413164736 Enamine

N-methoxy-N-methyl-2-(3,4,5-trifluorophenyl)cyclopropane-1-carboxamide

  • MDL number MFCD26935986
  • Cas Number : 1803609-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137066
Availability details for
Quantity
  • Availability
  • Add to Cart