ENA413164737 Enamine

3-(5-chloropyridine-2-carbonyl)-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD27920039
  • Cas Number : 1797776-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137170

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137170
Availability details for
Quantity
  • Availability
  • Add to Cart