ENA413164739 Enamine

5-(methylsulfanyl)-8-nitroquinoline

  • MDL number MFCD26935987
  • Cas Number : 1803591-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137202
Availability details for
Quantity
  • Availability
  • Add to Cart