ENA413164742 Enamine

tert-butyl N-[5-(hydroxyimino)-5,6,7,8-tetrahydronaphthalen-2-yl]carbamate

  • MDL number MFCD27500677
  • Cas Number : 1807938-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137717

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137717
Availability details for
Quantity
  • Availability
  • Add to Cart