ENA413164744 Enamine

2-{[5-(2-fluorophenoxymethyl)-1,2-oxazol-3-yl]methyl}-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD26935990
  • Cas Number : 1803591-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137845

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137845
Availability details for
Quantity
  • Availability
  • Add to Cart