ENA413164747 Enamine

2-({5-[(tert-butoxy)methyl]-1,2-oxazol-3-yl}methyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD27500680
  • Cas Number : 1803611-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137849
Availability details for
Quantity
  • Availability
  • Add to Cart