ENA413164748 Enamine

methyl 5-[(tert-butoxy)methyl]-1,2-oxazole-3-carboxylate

  • MDL number MFCD26935994
  • Cas Number : 1803583-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-137852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137852
Availability details for
Quantity
  • Availability
  • Add to Cart