ENA413164749 Enamine

tert-butyl 4-(carbamothioylamino)piperidine-1-carboxylate

  • MDL number MFCD26935995
  • Cas Number : 849546-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187...189
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137862

Safety Information

msds https://www.enaminestore.com/catalog/EN300-137862
Availability details for
Quantity
  • Availability
  • Add to Cart