ENA413164752 Enamine

4-bromo-1-(2,2,2-trifluoroethyl)-1H-imidazole

  • MDL number MFCD27920041
  • Cas Number : 1803608-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-137967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137967
Availability details for
Quantity
  • Availability
  • Add to Cart