ENA413164753 Enamine

pyridine-3-sulfonyl fluoride

  • MDL number MFCD26935998
  • Cas Number : 1373307-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138027

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138027
Availability details for
Quantity
  • Availability
  • Add to Cart