ENA413164756 Enamine

N-hydroxy-1-methyl-1H-pyrazole-4-carbonimidoyl chloride

  • MDL number MFCD26936002
  • Cas Number : 1282486-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138111
Availability details for
Quantity
  • Availability
  • Add to Cart