ENA413164757 Enamine

N-hydroxy-1,5-dimethyl-1H-pyrazole-4-carbonimidoyl chloride

  • Cas Number : 1704251-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138113
Availability details for
Quantity
  • Availability
  • Add to Cart