ENA413164759 Enamine

6-(4-chlorophenoxy)hexanamide

  • MDL number MFCD26936004
  • Cas Number : 1803587-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-138225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138225
Availability details for
Quantity
  • Availability
  • Add to Cart