ENA413164760 Enamine

methyl 6-[(2-fluorophenyl)methoxy]pyridine-3-carboxylate

  • MDL number MFCD26936005
  • Cas Number : 1276014-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138235

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138235
Availability details for
Quantity
  • Availability
  • Add to Cart