ENA413164761 Enamine

N-{[2-(phenoxymethyl)-1,3-thiazol-4-yl]methylidene}hydroxylamine

  • MDL number MFCD26936006
  • Cas Number : 1807938-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113...115
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138293

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138293
Availability details for
Quantity
  • Availability
  • Add to Cart