ENA413164762 Enamine

3-(1,3-dioxolan-2-yl)-1-[4-(trifluoromethyl)phenyl]piperidine

  • MDL number MFCD26936007
  • Cas Number : 1803587-09-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-138329
Availability details for
Quantity
  • Availability
  • Add to Cart