ENA413164775 Enamine

N'-(1-amino-2-ethoxy-2-oxoethylidene)amino 2-methoxyacetate

  • MDL number MFCD26936020
  • Cas Number : 1807939-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-138983

Safety Information

msds https://www.enaminestore.com/catalog/EN300-138983
Availability details for
Quantity
  • Availability
  • Add to Cart