ENA413164779 Enamine

5-(chloromethyl)-N,N-dimethyl-1,2,4-oxadiazol-3-amine

  • MDL number MFCD26936023
  • Cas Number : 1603533-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-139048

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139048
Availability details for
Quantity
  • Availability
  • Add to Cart