ENA413164783 Enamine

N,N-dimethyl-5-(piperidin-3-yl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD27920045
  • Cas Number : 1598686-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139053

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139053
Availability details for
Quantity
  • Availability
  • Add to Cart