ENA413164784 Enamine

N,N-dimethyl-5-(piperidin-4-yl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD27500689
  • Cas Number : 1594770-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-139054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139054
Availability details for
Quantity
  • Availability
  • Add to Cart