ENA413164785 Enamine

1-chloro-4-{[(chloromethyl)sulfanyl]methyl}benzene

  • MDL number MFCD27920046
  • Cas Number : 15894-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139103
Availability details for
Quantity
  • Availability
  • Add to Cart