ENA413164790 Enamine

4-(piperidin-3-yl)pyrrolidin-2-one

  • MDL number MFCD26936028
  • Cas Number : 1566966-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139165
Availability details for
Quantity
  • Availability
  • Add to Cart