ENA413164793 Enamine

5-methoxy-1,3-dimethyl-1H-pyrazole-4-sulfonamide

  • MDL number MFCD27920047
  • Cas Number : 88398-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139175

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139175
Availability details for
Quantity
  • Availability
  • Add to Cart