ENA413164796 Enamine

2-[6-(2,2,2-trifluoroethoxy)pyridin-3-yl]acetonitrile

  • MDL number MFCD27500690
  • Cas Number : 159981-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139202

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139202
Availability details for
Quantity
  • Availability
  • Add to Cart