ENA413164797 Enamine

sodium (diethylcarbamoyl)methanesulfonate

  • MDL number MFCD26936033
  • Cas Number : 1803585-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139206
Availability details for
Quantity
  • Availability
  • Add to Cart