ENA413164802 Enamine

methyl 2-butoxy-4-methoxybenzoate

  • MDL number MFCD26936038
  • Cas Number : 942132-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139264

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139264
Availability details for
Quantity
  • Availability
  • Add to Cart