ENA413164806 Enamine

3-(6-methylpyridine-3-carbonyl)-4H,5H,6H-cyclopenta[b]thiophen-2-amine

  • MDL number MFCD26936042
  • Cas Number : 1803610-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 206...208
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139350

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139350
Availability details for
Quantity
  • Availability
  • Add to Cart