ENA413164808 Enamine

3-benzyl 1-tert-butyl azetidine-1,3-dicarboxylate

  • MDL number MFCD26936044
  • Cas Number : 1803599-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139447

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139447
Availability details for
Quantity
  • Availability
  • Add to Cart