ENA413164812 Enamine

4-(dimethylamino)-N'-hydroxybutanimidamide

  • MDL number MFCD09940729
  • Cas Number : 1016814-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112339

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112339
Availability details for
Quantity
  • Availability
  • Add to Cart